1-(2-Pyridin-2-ylethyl)piperazine

Supplier: Matrix Scientific

033780-500MG
101836-354EA 307.69 USD
101836-354
1-(2-Pyridin-2-ylethyl)piperazine
1-(2-Pyridin-2-ylethyl)piperazine

Hazard: Irritant

Formula: C₁₁H₁₇N₃
MDL Number: MFCD01748645
CAS Number: 53345-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR