4-Furan-2-yl-4-oxobutyric acid

Supplier: Matrix Scientific

033778-500MG
101836-350EA 607.69 USD
101836-350
4-Furan-2-yl-4-oxobutyric acid
4-Furan-2-yl-4-oxobutyric acid

Hazard: Irritant

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00092710
CAS Number: 10564-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR