1-(3-Fluorobenzenesulfonyl)piperazine

Supplier: Matrix Scientific

033774-500MG
101836-342EA 307.69 USD
101836-342
1-(3-Fluorobenzenesulfonyl)piperazine
1-(3-Fluorobenzenesulfonyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₃FN₂O₂S
MDL Number: MFCD05262940
CAS Number: 743441-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR