(2,4-Dioxo-1,3-diazaspiro[4.4]non-3-yl)acetic acid

Supplier: Matrix Scientific

033773-500MG
101836-340EA 530.77 USD
101836-340
(2,4-Dioxo-1,3-diazaspiro[4.4]non-3-yl)acetic acid
(2,4-Dioxo-1,3-diazaspiro[4.4]non-3-yl)acetic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₄
MDL Number: MFCD05263199
CAS Number: 714-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR