3-Methyl-5-oxo-5H-thiazolo[3,2-a]pyrimidine-6-carboxylic acid
Supplier: Matrix Scientific
033746-500MG
101836-290EA
300
USD
101836-290
3-Methyl-5-oxo-5H-thiazolo[3,2-a]pyrimidine-6-carboxylic acid
3-Methyl-5-oxo-5H-thiazolo[3,2-a]pyrimidine-6-carboxylic acid
Hazard: Irritant
Formula:
C₈H₆N₂O₃S |
MDL Number:
MFCD03011581 CAS Number: 79932-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...