3-Morpholin-4-ylbenzoic acid

Supplier: Matrix Scientific

Synonyms: 3-(Morpholin-4-yl)benzoic acid3-Morpholinobenzoic acid4-(3-Carboxyphenyl)morpholine

033739-500MG
101836-276EA 307.69 USD
101836-276
3-Morpholin-4-ylbenzoic acid
3-Morpholin-4-ylbenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
MDL Number: MFCD06659078
CAS Number: 215309-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR