6-(3-Methoxyphenyl)imidazo[2,1-b]thiazole-5-carbaldehyde

Supplier: Matrix Scientific

033736-500MG
101836-270EA 307.69 USD
101836-270
6-(3-Methoxyphenyl)imidazo[2,1-b]thiazole-5-carbaldehyde
6-(3-Methoxyphenyl)imidazo[2,1-b]thiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₀N₂O₂S
MW: 258.3 g/mol
MDL Number: MFCD02661646
CAS Number: 439094-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR