4-Piperidin-1-ylbenzylamine

Supplier: Matrix Scientific

Synonyms: [4-(Piperidin-1-yl)phenyl]methylamine

033733-500MG
101836-264EA 32.31 USD
101836-264
4-Piperidin-1-ylbenzylamine
4-Piperidin-1-ylbenzylamine

Hazard: Irritant

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD06659080
CAS Number: 214759-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR