8-Chloro-2-methyl-quinoline-4-carboxylic acid

Supplier: Matrix Scientific

033722-500MG
101836-242EA 461.54 USD
101836-242
8-Chloro-2-methyl-quinoline-4-carboxylic acid
8-Chloro-2-methyl-quinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₈ClNO₂
MDL Number: MFCD00487565
CAS Number: 52413-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR