2-(6-Methylpyridin-3-yloxy)nicotinic acid

Supplier: Matrix Scientific

033717-500MG
101836-234EA 307.69 USD
101836-234
2-(6-Methylpyridin-3-yloxy)nicotinic acid
2-(6-Methylpyridin-3-yloxy)nicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₃
MDL Number: MFCD11053221
CAS Number: 1019107-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR