(1-Aminomethylcyclohexyl)dimethylamine

Supplier: Matrix Scientific

033711-500MG
101836-222EA 135.38 USD
101836-222
(1-Aminomethylcyclohexyl)dimethylamine
(1-Aminomethylcyclohexyl)dimethylamine

Hazard: Irritant

Formula: C₉H₂₀N₂
MDL Number: MFCD05201342
CAS Number: 41806-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR