2,5-Dimethyl-1-propyl-1H-pyrrole-3-carbaldehyde

Supplier: Matrix Scientific

033702-500MG
101836-204EA 307.69 USD
101836-204
2,5-Dimethyl-1-propyl-1H-pyrrole-3-carbaldehyde
2,5-Dimethyl-1-propyl-1H-pyrrole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₁₅NO
MDL Number: MFCD04624247
CAS Number: 18870-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR