3-(2-Fluorophenyl)-4,5-dihydroisoxazole-5-carboxylic acid

Supplier: Matrix Scientific

033670-500MG
101836-140EA 307.69 USD
101836-140
3-(2-Fluorophenyl)-4,5-dihydroisoxazole-5-carboxylic acid
3-(2-Fluorophenyl)-4,5-dihydroisoxazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈FNO₃
MDL Number: MFCD06011107
CAS Number: 842973-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR