3-(3-Chlorophenylmethanesulfonyl)propionic acid

Supplier: Matrix Scientific

033658-500MG
101836-116EA 307.69 USD
101836-116
3-(3-Chlorophenylmethanesulfonyl)propionic acid
3-(3-Chlorophenylmethanesulfonyl)propionic acid

Hazard: Irritant

Formula: C₁₀H₁₁ClO₄S
MDL Number: MFCD03830372
CAS Number: 702670-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR