1-(4-Fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-ylamine hydrochloride

Supplier: Matrix Scientific

033657-500MG
101836-114EA 307.69 USD
101836-114
1-(4-Fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-ylamine hydrochloride
1-(4-Fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-ylamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClFN₃
MDL Number: MFCD11506537
CAS Number: 1185304-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR