5-Propyl-isoxazole-3-carboxylic acid

Supplier: Matrix Scientific

033645-500MG
101836-090EA 461.54 USD
101836-090
5-Propyl-isoxazole-3-carboxylic acid
5-Propyl-isoxazole-3-carboxylic acid

Hazard: Irritant

Formula: C₇H₉NO₃
MW: 155.15 g/mol
MDL Number: MFCD05668697
CAS Number: 89776-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR