4-Hydroxy-2-oxo-1,2-dihydroquinoline-3-carboxylic acid
Supplier: Matrix Scientific
033640-5G
033640-1G
033640-500MG
101836-078EA
538.46
USD
101836-078
101836-076
101836-080
4-Hydroxy-2-oxo-1,2-dihydroquinoline-3-carboxylic acid
4-Hydroxy-2-oxo-1,2-dihydroquinoline-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₇NO₄ MW: 205.17 g/mol |
MDL Number:
MFCD06011091 CAS Number: 73776-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...