(3-Acetyl-2-methylindol-1-yl)acetic acid

Supplier: Matrix Scientific

033639-500MG
101836-074EA 307.69 USD
101836-074
(3-Acetyl-2-methylindol-1-yl)acetic acid
(3-Acetyl-2-methylindol-1-yl)acetic acid

Hazard: Irritant

Formula: C₁₃H₁₃NO₃
MDL Number: MFCD05844466
CAS Number: 717835-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR