N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-N-(3-chloro-2-methylphenyl)amine

Supplier: Matrix Scientific

031542-500MG
101831-910EA 306.15 USD
101831-910
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-N-(3-chloro-2-methylphenyl)amine
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-N-(3-chloro-2-methylphenyl)amine

Hazard: Irritant

Formula: C₁₇H₁₉Cl₂NO
MDL Number: MFCD10687623
CAS Number: 1040687-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR