2,5-Dimethyl-N-[3-(2-phenoxyethoxy)benzyl]aniline

Supplier: Matrix Scientific

031537-500MG
101831-900EA 306.15 USD
101831-900
2,5-Dimethyl-N-[3-(2-phenoxyethoxy)benzyl]aniline
2,5-Dimethyl-N-[3-(2-phenoxyethoxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₃H₂₅NO₂
MDL Number: MFCD10687619
CAS Number: 1040687-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR