N-[2-(4-Isopropylphenoxy)propyl]-2,5-dimethylaniline

Supplier: Matrix Scientific

031536-500MG
101831-898EA 306.15 USD
101831-898
N-[2-(4-Isopropylphenoxy)propyl]-2,5-dimethylaniline
N-[2-(4-Isopropylphenoxy)propyl]-2,5-dimethylaniline

Hazard: Irritant

Formula: C₂₀H₂₇NO
MDL Number: MFCD10687618
CAS Number: 1040687-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR