2,5-Dimethyl-N-(2-propoxybenzyl)aniline

Supplier: Matrix Scientific

031534-500MG
101831-894EA 306.15 USD
101831-894
2,5-Dimethyl-N-(2-propoxybenzyl)aniline
2,5-Dimethyl-N-(2-propoxybenzyl)aniline

Hazard: Irritant

Formula: C₁₈H₂₃NO
MDL Number: MFCD10687616
CAS Number: 1040687-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR