N-[2-(Isopentyloxy)benzyl]-2,5-dimethylaniline

Supplier: Matrix Scientific

031533-500MG
101831-892EA 306.15 USD
101831-892
N-[2-(Isopentyloxy)benzyl]-2,5-dimethylaniline
N-[2-(Isopentyloxy)benzyl]-2,5-dimethylaniline

Hazard: Irritant

Formula: C₂₀H₂₇NO
MDL Number: MFCD10687615
CAS Number: 1040687-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR