N-[2-(4-Methoxyphenoxy)propyl]-2,5-dimethylaniline

Supplier: Matrix Scientific

031531-500MG
101831-888EA 306.15 USD
101831-888
N-[2-(4-Methoxyphenoxy)propyl]-2,5-dimethylaniline
N-[2-(4-Methoxyphenoxy)propyl]-2,5-dimethylaniline

Hazard: Irritant

Formula: C₁₈H₂₃NO₂
MDL Number: MFCD10687613
CAS Number: 1040687-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR