N-(Cyclopropylmethyl)-2,5-dimethylaniline

Supplier: Matrix Scientific

031527-500MG
101831-880EA 306.15 USD
101831-880
N-(Cyclopropylmethyl)-2,5-dimethylaniline
N-(Cyclopropylmethyl)-2,5-dimethylaniline

Hazard: Irritant

Formula: C₁₂H₁₇N
MDL Number: MFCD03211310
CAS Number: 356539-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR