N-[4-(2-Methoxyethoxy)benzyl]-1-hexadecanamine

Supplier: Matrix Scientific

031524-500MG
101831-874EA 306.15 USD
101831-874
N-[4-(2-Methoxyethoxy)benzyl]-1-hexadecanamine
N-[4-(2-Methoxyethoxy)benzyl]-1-hexadecanamine

Hazard: Irritant

Formula: C₂₆H₄₇NO₂
MDL Number: MFCD10687608
CAS Number: 1040687-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR