N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-2,5-dimethylaniline

Supplier: Matrix Scientific

031522-500MG
101831-870EA 306.15 USD
101831-870
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-2,5-dimethylaniline
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-2,5-dimethylaniline

Hazard: Irritant

Formula: C₁₈H₂₂ClNO
MDL Number: MFCD10687606
CAS Number: 1040687-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR