3,5-Dimethyl-N-propylaniline

Supplier: Matrix Scientific

031521-500MG
101831-868EA 306.15 USD
101831-868
3,5-Dimethyl-N-propylaniline
3,5-Dimethyl-N-propylaniline

Hazard: Irritant

Formula: C₁₁H₁₇N
MDL Number: MFCD03210932
CAS Number: 13342-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR