3,5-Dimethyl-N-[2-(4-methylphenoxy)butyl]aniline
Supplier: Matrix Scientific
031513-500MG
101831-852EA
306.15
USD
101831-852
3,5-Dimethyl-N-[2-(4-methylphenoxy)butyl]aniline
3,5-Dimethyl-N-[2-(4-methylphenoxy)butyl]aniline
Hazard: Irritant
Formula:
C₁₉H₂₅NO |
MDL Number:
MFCD10687598 CAS Number: 1040686-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...