3,5-Dimethyl-N-(2-phenoxypropyl)aniline

Supplier: Matrix Scientific

031512-500MG
101831-850EA 306.15 USD
101831-850
3,5-Dimethyl-N-(2-phenoxypropyl)aniline
3,5-Dimethyl-N-(2-phenoxypropyl)aniline

Hazard: Irritant

Formula: C₁₇H₂₁NO
MDL Number: MFCD10687597
CAS Number: 1040686-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR