N-(4-Chlorobenzyl)-3,5-dimethylaniline

Supplier: Matrix Scientific

031506-500MG
101831-838EA 306.15 USD
101831-838
N-(4-Chlorobenzyl)-3,5-dimethylaniline
N-(4-Chlorobenzyl)-3,5-dimethylaniline

Hazard: Irritant

Formula: C₁₅H₁₆ClN
MDL Number: MFCD03210849
CAS Number: 356532-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR