3,5-Dimethyl-N-(2-methylbenzyl)aniline

Supplier: Matrix Scientific

031505-500MG
101831-836EA 306.15 USD
101831-836
3,5-Dimethyl-N-(2-methylbenzyl)aniline
3,5-Dimethyl-N-(2-methylbenzyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₉N
MDL Number: MFCD03210714
CAS Number: 356531-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR