3,5-Dichloro-N-[3-(2-phenoxyethoxy)benzyl]aniline

Supplier: Matrix Scientific

031502-500MG
101831-830EA 306.15 USD
101831-830
3,5-Dichloro-N-[3-(2-phenoxyethoxy)benzyl]aniline
3,5-Dichloro-N-[3-(2-phenoxyethoxy)benzyl]aniline

Hazard: Irritant

Formula: C₂₁H₁₉Cl₂NO₂
MDL Number: MFCD10687591
CAS Number: 1040686-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR