N-[2-(Isopentyloxy)benzyl]-1-hexadecanamine
Supplier: Matrix Scientific
031498-500MG
101831-822EA
306.15
USD
101831-822
N-[2-(Isopentyloxy)benzyl]-1-hexadecanamine
N-[2-(Isopentyloxy)benzyl]-1-hexadecanamine
Hazard: Irritant
Formula:
C₂₈H₅₁NO |
MDL Number:
MFCD10687587 CAS Number: 1040689-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...