3,5-Dichloro-N-(2-thienylmethyl)aniline

Supplier: Matrix Scientific

031497-500MG
101831-820EA 306.15 USD
101831-820
3,5-Dichloro-N-(2-thienylmethyl)aniline
3,5-Dichloro-N-(2-thienylmethyl)aniline

Hazard: Irritant

Formula: C₁₁H₉Cl₂NS
MDL Number: MFCD07579085
CAS Number: 870540-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR