3,5-Dichloro-N-[2-(2,6-dimethylphenoxy)ethyl]aniline

Supplier: Matrix Scientific

031496-500MG
101831-818EA 306.15 USD
101831-818
3,5-Dichloro-N-[2-(2,6-dimethylphenoxy)ethyl]aniline
3,5-Dichloro-N-[2-(2,6-dimethylphenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂NO
MW: 310.23 g/mol
MDL Number: MFCD10687586
CAS Number: 1040689-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR