N-(4-Butoxybenzyl)-1-hexadecanamine

Supplier: Matrix Scientific

031494-500MG
101831-814EA 306.15 USD
101831-814
N-(4-Butoxybenzyl)-1-hexadecanamine
N-(4-Butoxybenzyl)-1-hexadecanamine

Hazard: Irritant

Formula: C₂₇H₄₉NO
MDL Number: MFCD10687584
CAS Number: 1040689-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR