3,4-Dichloro-N-[2-(3-methylphenoxy)butyl]aniline

Supplier: Matrix Scientific

031490-500MG
101831-806EA 306.15 USD
101831-806
3,4-Dichloro-N-[2-(3-methylphenoxy)butyl]aniline
3,4-Dichloro-N-[2-(3-methylphenoxy)butyl]aniline

Hazard: Irritant

Formula: C₁₇H₁₉Cl₂NO
MDL Number: MFCD10687581
CAS Number: 1040688-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR