3,4-Dichloro-N-[2-(4-methoxyphenoxy)propyl]aniline

Supplier: Matrix Scientific

031489-500MG
101831-804EA 306.15 USD
101831-804
3,4-Dichloro-N-[2-(4-methoxyphenoxy)propyl]aniline
3,4-Dichloro-N-[2-(4-methoxyphenoxy)propyl]aniline

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂NO₂
MDL Number: MFCD10687580
CAS Number: 1040688-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR