3,4-Dichloro-N-[2-(4-chlorophenoxy)ethyl]aniline

Supplier: Matrix Scientific

031484-500MG
101831-794EA 306.15 USD
101831-794
3,4-Dichloro-N-[2-(4-chlorophenoxy)ethyl]aniline
3,4-Dichloro-N-[2-(4-chlorophenoxy)ethyl]aniline

Hazard: Irritant

Formula: C₁₄H₁₂Cl₃NO
MDL Number: MFCD03211062
CAS Number: 356537-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR