3,4-Dichloro-N-(2-furylmethyl)aniline

Supplier: Matrix Scientific

031483-500MG
101831-792EA 306.15 USD
101831-792
3,4-Dichloro-N-(2-furylmethyl)aniline
3,4-Dichloro-N-(2-furylmethyl)aniline

Hazard: Irritant

Formula: C₁₁H₉Cl₂NO
MDL Number: MFCD10687576
CAS Number: 51738-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR