3,4-Dichloro-N-(2-phenoxyethyl)aniline
Supplier: Matrix Scientific
031482-500MG
101831-790EA
306.15
USD
101831-790
3,4-Dichloro-N-(2-phenoxyethyl)aniline
3,4-Dichloro-N-(2-phenoxyethyl)aniline
Hazard: Irritant
Formula:
C₁₄H₁₃Cl₂NO |
MDL Number:
MFCD03210901 CAS Number: 356532-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...