N-(3-Chlorobenzyl)-1-hexadecanamine
Supplier: Matrix Scientific
031476-500MG
101831-778EA
306.15
USD
101831-778
N-(3-Chlorobenzyl)-1-hexadecanamine
N-(3-Chlorobenzyl)-1-hexadecanamine
Hazard: Irritant
Formula:
C₂₃H₄₀ClN |
MDL Number:
MFCD10687571 CAS Number: 1040688-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...