N-(Cyclohexylmethyl)-2,4,6-trimethylaniline
Supplier: Matrix Scientific
031473-500MG
101831-772EA
306.15
USD
101831-772
N-(Cyclohexylmethyl)-2,4,6-trimethylaniline
N-(Cyclohexylmethyl)-2,4,6-trimethylaniline
Hazard: Irritant
Formula:
C₁₆H₂₅N |
MDL Number:
MFCD03211363 CAS Number: 356540-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...