N-(2,4-Dichlorobenzyl)-2,4,6-trimethylaniline

Supplier: Matrix Scientific

031470-500MG
101831-766EA 306.15 USD
101831-766
N-(2,4-Dichlorobenzyl)-2,4,6-trimethylaniline
N-(2,4-Dichlorobenzyl)-2,4,6-trimethylaniline

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂N
MDL Number: MFCD03210795
CAS Number: 356531-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR