Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-2,4,6-trimethylaniline

Supplier: Matrix Scientific


031469-500MG
101831-764EA 260.23 USD InStock
101831-764
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-2,4,6-trimethylaniline
N-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]-2,4,6-trimethylaniline
CAS: 1040688-27-4
MDL Number: MFCD10687568
Molecular Formula: C19H24ClNO
Molecular Weight: 317,86
Hazard: Irritant


Formula: C₁₉H₂₄ClNO
MW: 317,86 g/mol
MDL Number: MFCD10687568
CAS Number: 1040688-27-4
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty