N-(4-Methoxyphenyl)-N-[2-(pentyloxy)benzyl]amine

Supplier: Matrix Scientific

031464-500MG
101831-754EA 306.15 USD
101831-754
N-(4-Methoxyphenyl)-N-[2-(pentyloxy)benzyl]amine
N-(4-Methoxyphenyl)-N-[2-(pentyloxy)benzyl]amine

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10687563
CAS Number: 1040688-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR