N-(4-Methoxyphenyl)-N-[4-(pentyloxy)benzyl]amine

Supplier: Matrix Scientific

031463-500MG
101831-752EA 306.15 USD
101831-752
N-(4-Methoxyphenyl)-N-[4-(pentyloxy)benzyl]amine
N-(4-Methoxyphenyl)-N-[4-(pentyloxy)benzyl]amine

Hazard: Irritant

Formula: C₁₉H₂₅NO₂
MDL Number: MFCD10687562
CAS Number: 1040687-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR