N-[2-(2,4-Dichlorophenoxy)ethyl]-4-methoxyaniline

Supplier: Matrix Scientific

031461-500MG
101831-748EA 306.15 USD
101831-748
N-[2-(2,4-Dichlorophenoxy)ethyl]-4-methoxyaniline
N-[2-(2,4-Dichlorophenoxy)ethyl]-4-methoxyaniline

Hazard: Irritant

Formula: C₁₅H₁₅Cl₂NO₂
MDL Number: MFCD03210944
CAS Number: 356532-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR