N-[4-(sec-Butoxy)benzyl]-4-phenoxyaniline

Supplier: Matrix Scientific

031458-500MG
101831-742EA 306.15 USD
101831-742
N-[4-(sec-Butoxy)benzyl]-4-phenoxyaniline
N-[4-(sec-Butoxy)benzyl]-4-phenoxyaniline

Hazard: Irritant

Formula: C₂₃H₂₅NO₂
MDL Number: MFCD10687560
CAS Number: 1040687-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR